SY3H3D679DE8 Synthonix Corporation

5-Chloro-1,3,4-thiadiazol-2-amine

  • MDL number MFCD01578280
  • Cas Number : 37566-40-8
  • Molecular Weight : 135.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-1,3,4-thiadiazol-2-amine
Inchi Code 1S/C2H2ClN3S/c3-1-5-6-2(4)7-1/h(H2,4,6)
Inchi Key OPGJORQBYBKWNH-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/39762.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39762.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart