SY3H3D679DF3 Synthonix Corporation

3-(Chloromethyl)-1H-1,2,4-triazol-5(4H)-one

  • MDL number MFCD07776630
  • Cas Number : 252742-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Pale-yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39844.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39844.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart