SY3H3D679DF6 Synthonix Corporation

O-(3-Chloroallyl)hydroxylamine

  • MDL number MFCD09951937
  • Cas Number : 87851-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39871.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39871.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart