SY3H3D679DFC Synthonix Corporation

3-(Carbamimidoylthio)propane-1-sulfonic acid

  • MDL number No MDL found
  • Cas Number : 21668-81-5
  • Molecular Weight : 198.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-{[amino(imino)methyl]sulfanyl}-1-propanesulfonic acid
Inchi Code 1S/C4H10N2O3S2/c5-4(6)10-2-1-3-11(7,8)9/h1-3H2,(H3,5,6)(H,7,8,9)
Inchi Key MQLJIOAPXLAGAP-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Refrigerator
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/39951.html

Safety Information

signalword Danger
pcodes P261
MSDS https://synthonix.com/39951.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart