SY3H3D679E04 Synthonix Corporation

4-Chloroisothiazole-5-carboxylic acid

  • Cas Number : 88982-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40015.html

Safety Information

MSDS https://synthonix.com/40015.html
Availability details for
Quantity
  • Availability
  • Add to Cart