SY3H3D679E0A Synthonix Corporation

5-Chloro-1H-pyrazole-4-carboxylic acid

  • Cas Number : 1186049-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/40033.html

Safety Information

MSDS https://synthonix.com/40033.html
Availability details for
Quantity
  • Availability
  • Add to Cart