SY3H3D679E7D Synthonix Corporation

2-Chloro-3-fluoro-5-nitropyridine

  • MDL number MFCD16659664
  • Cas Number : 1079179-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40475.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40475.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart