SY3H3D679E8E Synthonix Corporation

5-Chlorothiophene-2-carbonyl chloride

  • MDL number MFCD00043886
  • Cas Number : 42518-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40558.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40558.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart