SY3H3D679E8F Synthonix Corporation

4-Chloro-1H-pyrrole-2-carbonitrile

  • Cas Number : 57097-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40566.html

Safety Information

MSDS https://synthonix.com/40566.html
Availability details for
Quantity
  • Availability
  • Add to Cart