SY3H3D679E9A Synthonix Corporation

3-Chloro-2-hydroxy-5-nitropyridine

  • MDL number MFCD06656618
  • Cas Number : 22353-38-4
  • Molecular Weight : 174.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: Off white - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40599.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40599.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart