SY3H3D679EA6 Synthonix Corporation

4-Chloro-2-fluoropyridine

  • MDL number MFCD04112503
  • Cas Number : 34941-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40643.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40643.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart