SY3H3D679EA8 Synthonix Corporation

3-Chloro-4-Iodopyridine

  • MDL number MFCD00272199
  • Cas Number : 77332-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40645.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/40645.html
pictograms GHS07
hstatements H302, H312, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart