SY3H3D679ED3 Synthonix Corporation

3-Chloro-2-hydroxypyridine

  • MDL number MFCD01646109
  • Cas Number : 13466-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Gray - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40847.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/40847.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart