SY3H3D679EDE Synthonix Corporation

4-Chloropyridine-3-sulfonamide

  • MDL number MFCD04038136
  • Cas Number : 33263-43-3
  • Molecular Weight : 192.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40904.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40904.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart