SY3H3D679EE5 Synthonix Corporation

2-(Chloromethyl)pyrimidin-4(3H)-one

  • Cas Number : 944905-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40934.html

Safety Information

MSDS https://synthonix.com/40934.html
Availability details for
Quantity
  • Availability
  • Add to Cart