SY3H3D679EF0 Synthonix Corporation

1-Cyanocyclopropanecarboxamide

  • Cas Number : 1559-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41029.html

Safety Information

MSDS https://synthonix.com/41029.html
Availability details for
Quantity
  • Availability
  • Add to Cart