SY3H3D679EFA Synthonix Corporation

5-Chloro-2-methylpyrimidin-4-amine

  • Cas Number : 438249-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41080.html

Safety Information

MSDS https://synthonix.com/41080.html
Availability details for
Quantity
  • Availability
  • Add to Cart