SY3H3D679F01 Synthonix Corporation

2-(Chloromethyl)-1-methyl-1H-imidazole hydrochloride

  • MDL number MFCD00234118
  • Cas Number : 78667-04-6
  • Molecular Weight : 167.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41132.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41132.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart