SY3H3D679F52 Synthonix Corporation

6-Chloro-N3-methylpyridazine-3,4-diamine

  • Cas Number : 17645-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41161.html

Safety Information

MSDS https://synthonix.com/41161.html
Availability details for
Quantity
  • Availability
  • Add to Cart