SY3H3D679F6A Synthonix Corporation

4-Chloro-N-methoxy-N-methylbutanamide

  • Cas Number : 64214-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41545.html

Safety Information

MSDS https://synthonix.com/41545.html
Availability details for
Quantity
  • Availability
  • Add to Cart