SY3H3D679F6C Synthonix Corporation

4-Chloro-1,1-dimethoxybutane

  • MDL number MFCD09032953
  • Cas Number : 29882-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41597.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41597.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart