SY3H3D679F71 Synthonix Corporation

2-Chloro-4-fluoronicotinonitrile

  • Cas Number : 1054552-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41683.html

Safety Information

MSDS https://synthonix.com/41683.html
Availability details for
Quantity
  • Availability
  • Add to Cart