SY3H3D679F73 Synthonix Corporation

2-Chloro-3-iodoisonicotinonitrile

  • Cas Number : 908279-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41685.html

Safety Information

MSDS https://synthonix.com/41685.html
Availability details for
Quantity
  • Availability
  • Add to Cart