SY3H3D679F75 Synthonix Corporation

2-Chloro-5-nitro-3-(trifluoromethyl)pyridine

  • MDL number No MDL found
  • Cas Number : 99368-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41693.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/41693.html
pictograms GHS05, GHS06
hstatements H301, H315, H318, H335
Availability details for
Quantity
  • Availability
  • Add to Cart