SY3H3D679F77 Synthonix Corporation

2-Chloro-5-iodo-3-(trifluoromethyl)pyridine

  • MDL number MFCD12024302
  • Cas Number : 887707-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41714.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41714.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart