SY3H3D679F7B Synthonix Corporation

2-Chloro-3,4-difluoronitrobenzene

  • Cas Number : 169468-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/41748.html

Safety Information

MSDS https://synthonix.com/41748.html
Availability details for
Quantity
  • Availability
  • Add to Cart