SY3H3D679F7C Synthonix Corporation

2-Chloro-3,5-difluoronitrobenzene

  • Cas Number : 36556-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41749.html

Safety Information

MSDS https://synthonix.com/41749.html
Availability details for
Quantity
  • Availability
  • Add to Cart