SY3H3D679F84 Synthonix Corporation

5-Chloro-2,4-difluorobenzenesulfonylchloride

  • MDL number No MDL found
  • Cas Number : 13656-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41763.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41763.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart