SY3H3D679F8A Synthonix Corporation

4-Chloro-3-fluoroiodobenzene

  • MDL number MFCD00042580
  • Cas Number : 202982-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41807.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41807.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart