SY3H3D679F94 Synthonix Corporation

7-Chlorothieno[3,2-d]pyrimidine

  • Cas Number : 31492-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/41832.html

Safety Information

MSDS https://synthonix.com/41832.html
Availability details for
Quantity
  • Availability
  • Add to Cart