SY3H3D67A0ED Synthonix Corporation

5-Chloro-4-fluoro-2-nitrophenol

  • Cas Number : 65001-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/41976.html

Safety Information

MSDS https://synthonix.com/41976.html
Availability details for
Quantity
  • Availability
  • Add to Cart