SY3H3D67A10C Synthonix Corporation

2-Chloro-4-iodo-6-nitroaniline

  • MDL number MFCD26936371
  • Cas Number : 873386-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Light-yellow to yellow or orange powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42107.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42107.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart