SY3H3D67A113 Synthonix Corporation

7-Chloro-1H-benzo[d][1,2,3]triazole

  • Cas Number : 67130-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42159.html

Safety Information

MSDS https://synthonix.com/42159.html
Availability details for
Quantity
  • Availability
  • Add to Cart