SY3H3D67A119 Synthonix Corporation

2-Chloro-5-nitronicotinamide

  • MDL number MFCD01860226
  • Cas Number : 60524-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42171.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42171.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart