SY3H3D67A11E Synthonix Corporation

6-Chloro-1H-imidazo[4,5-c]pyridin-2(3H)-one

  • Cas Number : 7205-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42180.html

Safety Information

MSDS https://synthonix.com/42180.html
Availability details for
Quantity
  • Availability
  • Add to Cart