SY3H3D67A120 Synthonix Corporation

3,5-Dibromo-2-chloro-4-methylpyridine

  • MDL number MFCD09864796
  • Cas Number : 1000017-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to pale-yellow solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42187.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42187.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart