SY3H3D67A124 Synthonix Corporation

3-Chloro-2,4-difluoroaniline

  • MDL number MFCD00042200
  • Cas Number : 2613-34-5
  • Molecular Weight : 163.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-2,4-difluorophenylamine
Inchi Code 1S/C6H4ClF2N/c7-5-3(8)1-2-4(10)6(5)9/h1-2H,10H2
Inchi Key BNTNWQPIBPBJOO-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Refrigerator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/42235.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42235.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart