SY3H3D67A126 Synthonix Corporation

4-Chloro-2,6-difluoroaniline

  • Cas Number : 69411-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42237.html

Safety Information

MSDS https://synthonix.com/42237.html
Availability details for
Quantity
  • Availability
  • Add to Cart