SY3H3D67A128 Synthonix Corporation

4-Chloro-2-fluoro-6-iodoaniline

  • MDL number MFCD01318122
  • Cas Number : 216393-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Yellow to light-brown to dark-brown or purple powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42239.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42239.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart