SY3H3D67A12E Synthonix Corporation

2-Chloro-5-nitrophenol

  • MDL number MFCD01571825
  • Cas Number : 619-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Pale yellow - Greyish reddish yellow - Yellow-brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42266.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42266.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart