SY3H3D67A136 Synthonix Corporation

4-Chloro-6-hydroxynicotinic acid

  • Cas Number : 73038-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/42274.html

Safety Information

MSDS https://synthonix.com/42274.html
Availability details for
Quantity
  • Availability
  • Add to Cart