SY3H3D67A13B Synthonix Corporation

4-Chloro-3-nitrobenzenesulfonic acid

  • MDL number MFCD00274152
  • Cas Number : 121-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42289.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42289.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart