SY3H3D67A149 Synthonix Corporation

5-Chloro-2-fluorophenol

  • MDL number MFCD04116357
  • Cas Number : 186589-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42358.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42358.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart