SY3H3D67A14E Synthonix Corporation

5-Chloro-2-iodophenol

  • MDL number MFCD09753473
  • Cas Number : 136808-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42363.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42363.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart