SY3H3D67A152 Synthonix Corporation

3-Chloro-4-nitroaniline

  • MDL number MFCD00085922
  • Cas Number : 825-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Pale yellow - Yellow red
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42400.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42400.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart