SY3H3D67A157 Synthonix Corporation

3-Chloro-6-methylpyrazine-2-carboxylic acid

  • MDL number MFCD22481007
  • Cas Number : 1384678-93-6
  • Molecular Weight : 172.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42409.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42409.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart