SY3H3D67A15D Synthonix Corporation

2-Chloro-5-methoxy-3-nitropyridine

  • Cas Number : 1003711-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42437.html

Safety Information

MSDS https://synthonix.com/42437.html
Availability details for
Quantity
  • Availability
  • Add to Cart