SY3H3D67A16D Synthonix Corporation

7-Chloro-1-methyl-1H-pyrazolo[4,3-d]pyrimidine

  • MDL number No MDL found
  • Cas Number : 923282-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42500.html

Safety Information

MSDS https://synthonix.com/42500.html
Availability details for
Quantity
  • Availability
  • Add to Cart