SY3H3D67A17B Synthonix Corporation

3-Chloro-5-fluoroaniline

  • MDL number MFCD03407962
  • Cas Number : 4863-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Yellow red - Brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42568.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42568.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart