SY3H3D67A17C Synthonix Corporation

2-Chloro-5-fluoro-3-picoline

  • MDL number MFCD04972725
  • Cas Number : 38186-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Lump / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42569.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42569.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart